missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ PU900 Series Medium Duty Polyurethane Gloves

Provides good mechanical resistance to abrasion and cuts. Ansell PU900 Series Medium Duty Polyurethane Gloves provide comfort and dexterity.

£185.00

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003
Description Gauge: 13
Color White, Gray
Exterior Finish Palm Coated
Material Polyurethane
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15630934
View Documents
Ansell™
B090100
10 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10558645
View Documents
Ansell™
B090070
7 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10660956
View Documents
Ansell™
B090080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15640934
View Documents
Ansell™
B090090
9 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Mechanical protection gloves
  • Medium duty cut resistance gloves
  • Good abrasion and cut resistance
  • Low lint glove
  • Good comfort and dexterity

Applications:

  • Assembly of white goods
  • Plastics injection and moulding
  • Assembling metal parts and components
  • Aeronautics
  • Finishing works
Specifications

Specifications

EN 420:2003 + A1:2009 and EN 388:2003
White, Gray
Polyurethane
Construction: Knitted
Spandex Dyneema™
PU900
Gauge: 13
Palm Coated
Mechanical Protection Gloves
Knit
12 pairs in a bag, 240 pairs in a carton
196 to 252 mm
Videos
SDS
Documents

Documents

Product Certifications