Learn More
SVL™ Rotulex™ PTFE O-ring for Spherical Joint
Offers a 'grease-free' spherical joint. Rotulex™ PTFE O-ring for Spherical Joint is highly resistant to chemical corrosion. Can be used at temperatures up to 200 °C. PTFE covered O-ring fits into a female cup joint which can be retained together with spring loaded metal clip.
Brand:  SVL RO64
 View more versions of this product
Product Code. 12009032
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Offers a grease-free spherical joint
  • PTFE pathway is highly resistant to chemical corrosion
  • Can be used at temperatures up to 200°C
  • PTFE covered O-ring fits into a female cup joint
  • Joint can be retained together with spring loaded metal clip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
40 mm
45 mm Shank
64 mm
RM:RF64 Glass Joint
1/Pk.
PTFE O-ring