missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Proteinase K
Delivered with a stabilizing solution for enhanced stability. Macherey-Nagel™ Proteinase K is lyophilized proteinase K supplied in pre-measured vials for easy preparation of 20mg/mL stock solution. It can be used with NucleoSpin™ and NucleoBond™ kits.
£40.20 - £170.00
Specifications
Concentration | 20 mg/mL |
---|---|
Color | White |
Components | Buffer solution |
For Use With (Application) | Viral RNA and DNA isolation, including: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1 |
For Use With (Equipment) | NucleoSpin™ and NucleoBond™ kits |
Product Code | Brand | Quantity | Storage Buffer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Storage Buffer | Price | Quantity & Availability | ||||
15880918
|
Macherey-Nagel™
740506.30 |
30 mg | 1.8mL Proteinase Buffer PB |
£40.20
30mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11912312
|
Macherey-Nagel™
740506 |
100 mg (2 x 50 mg) | 8mL Proteinase Buffer PB |
£170.00
100mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Proteinase K is necessary for the isolation of DNA or simultaneous RNA / DNA isolation for DNA viruses (e.g. HBV) that are more difficult to lyse. Proteinase K treatment is also recommended for viral RNA isolation when viscous samples have to be processed (e.g., sputum samples).
- Delivered with stabilizing solution to enable storage for up to six months at −20°C
- No decrease of proteinase activity when dissolved in the buffer compared to dissolving in water
- Lyophilized mataerial is supplied in precisely measured vials, to which buffer solution is added to prepare 20mg/mL stock solution
- Suitable for use with the NucleoSpin™ RNA Virus F kit and other NucleoSpin™ and NucleoBond™ kits, and also suitable for use in most other applications
Specifications
20 mg/mL | |
Buffer solution | |
NucleoSpin™ and NucleoBond™ kits | |
Proteinase and Buffer, separately: room temperature (18–25°C) for at least 1 year Prepared Stock Solution: −20°C for up to 6 months |
|
Proteinase K |
White | |
Viral RNA and DNA isolation, including: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1 | |
Vial | |
Tritirachium album | |
Lyophilized |