missing translation for 'onlineSavingsMsg'
Learn More

Propargyl acetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7045260
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-09-8
C5H6O2
98.101
MFCD00041601
RIZZXCJMFIGMON-UHFFFAOYSA-N
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
69388
prop-2-ynyl acetate
CC(=O)OCC#C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-09-8
C5H6O2
98.101
MFCD00041601
RIZZXCJMFIGMON-UHFFFAOYSA-N
propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
69388
prop-2-ynyl acetate
CC(=O)OCC#C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.989
Boiling Point 124°C
Flash Point 31°C (88°F)
Odor Characteristic
Linear Formula CH3CO2CH2C≡CH
Refractive Index 1.4185
Quantity 5 g
UN Number UN1992
Beilstein 1742046
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material Propargyl acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'