missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PRM1 |
---|---|
Concentration | 553 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894984
|
Proteintech
15697-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884984
|
Proteintech
15697-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PRM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRM1. This antibody recognizes human, mouse, rat antigen. The PRM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PRM1 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
553 μg/mL | |
Polyclonal | |
Rabbit | |
5619 | |
Primary |