missing translation for 'onlineSavingsMsg'
Learn More

PRKCG (PKC gamma) Recombinant Human Protein

PRKCG (PKC gamma) Protein

POA

Specifications

Accession Number NP_002730
Common Name PRKCG (PKC gamma) Recombinant Human Protein
Concentration 0.4mg/mL
Expression System Baculovirus
Gene Alias PRKCG (PKC gamma)
View More Specs

Description

Description

The PKC family of serine⁄threonine kinases, including PRKCG (PKC gamma), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications

Specifications

NP_002730
0.4mg/mL
PRKCG (PKC gamma)
1 Each
95% by SDS-PAGE
1800nmoles of phosphate transferred to histone H1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.5 to 2μg/mL
Kinase assay
None
PRKCG (PKC gamma) Recombinant Human Protein
Baculovirus
PRKCG (PKC gamma) Recombinant Human Protein
PRKCG (PKC gamma) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
–80°C
Human
SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.