missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PPP3CC |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836645
|
Proteintech
55163-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16826645
|
Proteintech
55163-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to PPP3CC.
Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca, calcineurin A-beta, and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).Specifications
PPP3CC | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P48454 | |
19057, 5533 | |
Peptide | |
Primary | |
-20°C | |
PPP3CC |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
36 53aa, CALNA3, CAM PRP catalytic subunit, CNA3, PPP3CC | |
Ppp3cc | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |