missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Powercoat™ 080-10 Butyl™ Gloves

Product Code. 12630963
Click to view available options
Size: 9
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect yourself against aggressive chemicals, acetones, MEK and esters. Honeywell Safety Products™ Powercoat™ 080-10 Butyl Gloves are recommended for offshore, petro-chemical, and water treatment industries.

  • Rolled edge stops drops and reinforces tear resistance.
  • Smooth inside to ease donning and duffing.
  • More flexible than natural latex.
  • Good dexterity
  • Resistant to many solvents, acids and medium concentrated bases.
  • Resists ozone and heat.
  • Gaz tight.
  • Good tear resistance.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE 3, Quality Assurance ISO 9001/2000, EC Attestation Number 0075/797/162/05/13/0411
Classification Dexterity: 5
Color Black
Cut Resistance Rating 1
Material Butyl
Product Type Gloves
Size 9
Thickness (Metric) 0.5 mm
For Use With (Application) Offshore, Petrochemical and Water Treatment industries
Glove Cuff Style Rolled
Puncture Rating 1
Resistant To Aggressive chemicals, acetones, MEK and esters
Tear Rating 1
Length (Metric) 35 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'