missing translation for 'onlineSavingsMsg'
Learn More

Potassium selenate, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11430950
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-59-2
Molecular Formula K2O4Se
Molecular Weight (g/mol) 221.164
MDL Number MFCD00054125
InChI Key YAZJAPBTUDGMKO-UHFFFAOYSA-L
Synonym potassium selenate, dipotassium selenate, unii-99798pu7mi, selenic acid, dipotassium salt, 2k.so4e, selenic acid solution, selenic acid, potassium salt 1:2, selenic acid solution, 40 wt. % in water 100g
PubChem CID 62680
IUPAC Name dipotassium;selenate
SMILES [O-][Se](=O)(=O)[O-].[K+].[K+]

Specifications

Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water
Formula Weight 221.16
Odor Odorless
Sensitivity Hygroscopic
Density 3.07
Chemical Name or Material Potassium selenate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'