Learn More
Potassium peroxydisulfate, Electrophoresis Grade, 99.0%, Thermo Scientific Chemicals
£10.65 - £29.55
Chemical Identifiers
CAS | 7727-21-1 |
---|---|
Molecular Formula | K2O8S2 |
Molecular Weight (g/mol) | 270.309 |
MDL Number | MFCD00011386 |
InChI Key | USHAGKDGDHPEEY-UHFFFAOYSA-L |
Synonym | potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc |
PubChem CID | 24412 |
IUPAC Name | dipotassium;sulfonatooxy sulfate |
SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420627
|
Thermo Scientific Alfa Aesar
J66672.06 |
5 g | N/A | ||||||
15430627
|
Thermo Scientific Alfa Aesar
J66672.14 |
25 g |
£18.69
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440627
|
Thermo Scientific Alfa Aesar
J66672.22 |
100 g |
£29.55
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7727-21-1 | |
270.309 | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
24412 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] |
K2O8S2 | |
MFCD00011386 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
dipotassium;sulfonatooxy sulfate |
Specifications
100°C (decomposition) | |
White | |
K2O8S2 | |
UN1492 | |
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+] | |
270.309 | |
270.31 | |
Electrophoresis | |
2.477g/mL |
7727-21-1 | |
Crystalline | |
MFCD00011386 | |
14,7656 | |
USHAGKDGDHPEEY-UHFFFAOYSA-L | |
dipotassium;sulfonatooxy sulfate | |
24412 | |
99.0% | |
Odorless | |
Potassium peroxydisulfate |
Safety and Handling
GHS H Statement
H334-H272-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May intensify fire
oxidizer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
P221-P210-P285-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of inadequate ventilation wear respiratory protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: POTASSIUM PERSULFATE
missing translation for 'rtecsNumber' : SE0400000
missing translation for 'tsca' : TSCA
RUO – Research Use Only