missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals

£48.84 - £94.67

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347678
View Documents Promotion Details
Thermo Scientific Alfa Aesar
033351.30
250 g
£48.84
250g
Please to purchase this item. Need a web account? Register with us today!
11337678
View Documents
Thermo Scientific Alfa Aesar
033351.A1
1 kg
£94.67
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

As a primary standard for preparing volumetric alkali solutions and as a buffer in pH determinations. Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters. It acts as a buffer in pH determination. It finds application as a useful standard for total organic carbon testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

877-24-7
204.222
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
MFCD00013070
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
potassium;2-carboxybenzoate

Specifications

297°C (decomposition)
White
99.95-100.05%
MFCD00013070
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
23676735
ACS Reagent
Potassium hydrogen phthalate, primary standard, For ACS analysis
877-24-7
Crystalline Powder
C8H5KO4
3637128
Soluble in water. Slightly soluble in alcohol.
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
204.23
1.636 g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 212-889-4

missing translation for 'rtecsNumber' : CZ4326000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only