missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, ≥99.5%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 398°c |
Color | Orange |
pH | 3.5 (at 20°C) |
Quantity | 25 g |
Linear Formula | K2Cr2O7 |
UN Number | UN3086 |
Formula Weight | 294.18g/mol |
Percent Purity | ≥99.5% |
Odor | Odorless |
Packaging | Poly bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'