missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, ≥99.5%, Honeywell™, BioXtra
BioXtra,≥99.5%
51.06€ - 4271.35€
Chemical Identifiers
CAS | 7778-50-9 |
---|---|
Molecular Formula | Cr2K2O7 |
Molecular Weight (g/mol) | 294.182 |
MDL Number | MFCD00011367 |
InChI Key | KMUONIBRACKNSN-UHFFFAOYSA-N |
Synonym | potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
PubChem CID | 24502 |
ChEBI | CHEBI:53444 |
IUPAC Name | dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Description
- Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
- Includes application-specific formulations, such as tablets and prepared solutions.
Chemical Identifiers
7778-50-9 | |
294.182 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
24502 | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Cr2K2O7 | |
MFCD00011367 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
CHEBI:53444 | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Specifications
398°C | |
Orange | |
Crystalline | |
K2Cr2O7 | |
UN3086 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24502 | |
294.18g/mol | |
Odorless | |
Potassium dichromate |
7778-50-9 | |
3.5 (at 20°C) | |
Cr2K2O7 | |
MFCD00011367 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
294.182 | |
CHEBI:53444 | |
≥99.5% | |
2.7g/cu.cm at 20°C |
Safety and Handling
Missing Phrase - N15.00950417-P201-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P313
missing translation for 'dotInformation' : class 6.1 - PG 2 - EHS - RQ -Toxic solids, oxidizing, n.o.s., HI: all
missing translation for 'rtecsNumber' : 231-906-6