missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M Potassium Chloride with AgCl, Electrode Filling Solution, Reagecon™
Used for refilling or replacing the reference electrolyte in silver/silver chloride or calomel reference electrodes. 3M Potassium Chloride with AgCl, Electrode Filling Solution, Reagecon™ is the filling solution of choice in either potentiometric titrations or direct pH, redox or ion measurements.
£70.95 - £87.35
Chemical Identifiers
CAS | 7447-40-7, 7783-90-6 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11593162
|
Reagecon
EFS3AC |
100 mL |
£70.95
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11503172
|
Reagecon
EFS3AC5 |
500 mL |
£87.35
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A good electrolyte must fulfil a number of conditions: the equitransference of the cation/anion combination should be as close as possible to being equimobile, have constant chloride activity, be of high electrical conductance and as non-chemically reactive as possible.
Potassium Chloride (KCl) fulfils all of these conditions to a greater or lesser extent, and is the filling solution of choice in either potentiometric titrations or direct pH, redox or ion measurements where silver/silver chloride or calomel reference electrodes are used. 3M KCl is appropriate where the working temperature is at least 20°C.
Chemical Identifiers
7447-40-7, 7783-90-6 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
CHEBI:32588 |
ClK | |
MFCD00011360 | |
4873 | |
[Cl-].[K+] |
Specifications
7447-40-7, 7783-90-6 | |
Liquid | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
4873 |
3 mol/L, Saturated with AgCl | |
Electrode Filling | |
MFCD00011360 | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 |