missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 292°C |
Color | White |
pH | 7.0 to 8.0 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 98.14g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'