missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POP7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14964-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. Also a component of RNase MRP complex, which cleaves pre-rRNA sequences.Specifications
POP7 | |
Polyclonal | |
Unconjugated | |
POP7 | |
0610037N12Rik, hPOP7, POP7, RNaseP protein p20, RPP2, RPP20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10248 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75817 | |
POP7 | |
POP7 Fusion Protein Ag6827 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |