Learn More
Corning™ Polystyrene Stripette Pipets
Manufactured with optically clear polystyrene for accurate measurement. Corning Polystyrene Stripette™ Pipets are ideal for sterile tissue culture applications.
POA
Specifications
Material Polystyrene
Type Stripette
View More Specs
Description
Description
  • Triple bagged
  • Without plug
  • Sterile
Specifications
Specifications
Polystyrene
Stripette
SDS