missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Sanitary Eccentric Reducer

Maintain sanitary conditions in your processes

Brand:  Masterflex™ 31805-01

Product Code. 11769579

  • £240.00 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Meet USP Class VI for pharmaceutical use and FDA, USDA and 3A standards for food contact.
  • All fittings are compatible with Tri-Clamp™; fittings.
  • Excellent chemical resistance.
  • Low leachable characteristics.
Specifications

Specifications

PP Sanitary Eccentric Reducer, 1 1/2 in. x 3/4 in. Mini Clamp Size
Polypropylene
-9°C
1 1/2 in., 3/4 in.
125 psi in ambient conditions, using plastic true union style clamp
82°F
Sanitary Connection
15°F
Very good chemical resistance
Autoclave or Ethylene Oxide
180°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers