missing translation for 'onlineSavingsMsg'
Learn More

Platin-Iridium-Draht, 0.2 mm (0.008 in.) Durchmesser, > 99.9 %, Thermo Scientific Chemicals

Form: Ca. 0.7 g/10 cm; Härte: weich; Formel: Pt:Ir; 80:20 wt%

142.00 CHF - 528.00 CHF

Chemische Identifikatoren

CAS 7440-06-4
Summenformel Pt
Molekulargewicht (g/mol) 195.08
MDL-Nummer MFCD00011179
InChI-Schlüssel BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC-Name Platin
SMILES [Pt]
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
11348328
Unterlagen ansehen Aktionsdetails
Thermo Scientific Alfa Aesar
039526.BS
10 cm
142.00 CHF
10 Zentimeter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
11338328
Unterlagen ansehen
Thermo Scientific Alfa Aesar
039526.BW
50 cm
528.00 CHF
50 Zentimeter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
Platin
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Spezifikation

Platinum Iridium
7440-06-4
>99.9%
MFCD00011179
Insoluble in water and acid.
[Pt]
195.08
CHEBI:33400
Draht, 0.2 mm (0.008 Zoll) Durchmesser
Silberfarben bis grau
Silver gray wire
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
Platin
23939
Geruchsneutral
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

missing translation for 'tsca' : Ja

Recommended Storage : Umgebungstemperaturen

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

RUO – Research Use Only