missing translation for 'onlineSavingsMsg'
Learn More

Platingewebe, Unimesh N7433, erweitertes Metallgewebe, 0.34 mm (0.013 in.) Dicke, Thermo Scientific Chemicals

457.00 CHF - 1725.00 CHF

Chemical Identifiers

CAS 7440-06-4
Summenformel Pt
Molekulargewicht (g/mol) 195.08
MDL-Nummer MFCD00011179
InChI-Schlüssel BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC-Name Platin
SMILES [Pt]
View More Specs

Products 2
Product Code Brand Menge Price Quantity & Availability  
Product Code Brand Menge Price Quantity & Availability  
15427355
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
046966.FF
25 x 25 mm
457.00 CHF
1 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Add to basket
 
15437355
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
046966.FI
50 x 50 mm
1725.00 CHF
1 Stück
Verfügbar ab: 20-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Add to basket
 
Description

Description

Platinum gauze is used in electronics industry for electrical contacts which might be subjected to high temperatures. It also acts as a catalyst in the production of nitric acid. It plays an essential role in the cracking processes in oil refineries. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
Platin
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

Platinum gauze, Unimesh N7433
7440-06-4
MFCD00011179
Soluble in hot aqua regia.
[Pt]
195.08
CHEBI:33400
Geruchsneutral
Expanded metal mesh, 0.34mm (0.013 in.) thick
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
Platin
23939
0.34mm Mesh
≈1.0g/25x25mm
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-116-1

missing translation for 'rtecsNumber' : TP2160000

missing translation for 'tsca' : Yes

Recommended Storage : Umgebungstemperaturen

SDS
Documents

Documents

Certificates

RUO – Research Use Only