missing translation for 'onlineSavingsMsg'
Learn More

Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals

£25.33 - £645.21

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10013863
View Documents Promotion Details
Thermo Scientific Acros
195240010
1 g Glass bottle
£25.33
1g
Please to purchase this item. Need a web account? Register with us today!
10552601
View Documents
Thermo Scientific Acros
195240100
10 g Glass bottle
£130.65
10g
Please to purchase this item. Need a web account? Register with us today!
10562601
View Documents
Thermo Scientific Acros
195240500
50 g Glass bottle
£645.21
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

Black
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
10% (Pt)
Platinum on activated carbon
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
195.08
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Self-heating in large quantities; may catch fire.

GHS P Statement:
Keep cool. Protect from sunlight.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Platinum on activated carbon, 10% Pt, ca. 50% moisture

SDS
Documents

Documents

Certificates

RUO – Research Use Only