missing translation for 'onlineSavingsMsg'
Learn More

piperidine-1-carboximidamide hydroiodide, Thermo Scientific™

£86.30 - £171.00

Chemical Identifiers

CAS 102392-91-6
Molecular Formula C6H14IN3
Molecular Weight (g/mol) 255.103
InChI Key MKDZAWNSOJLXKY-UHFFFAOYSA-N
Synonym piperidine-1-carboximidamide hydroiodide, piperidinecarboxamidine, iodide, c6h13n3.hi
PubChem CID 2778397
IUPAC Name piperidine-1-carboximidamide;hydroiodide
SMILES C1CCN(CC1)C(=N)N.I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10055132
View Documents
Thermo Scientific Maybridge
KM00642DA
1 g
£86.30
1g
Please to purchase this item. Need a web account? Register with us today!
10649905
View Documents
Thermo Scientific Maybridge
5 g
£171.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

102392-91-6
255.103
piperidine-1-carboximidamide hydroiodide, piperidinecarboxamidine, iodide, c6h13n3.hi
piperidine-1-carboximidamide;hydroiodide
C6H14IN3
MKDZAWNSOJLXKY-UHFFFAOYSA-N
2778397
C1CCN(CC1)C(=N)N.I

Specifications

102392-91-6
C6H14IN3
MKDZAWNSOJLXKY-UHFFFAOYSA-N
piperidine-1-carboximidamide;hydroiodide
2778397
97%
97%
piperidine-1-carboximidamide hydroiodide, piperidinecarboxamidine, iodide, c6h13n3.hi
C1CCN(CC1)C(=N)N.I
255.103
255.1
Piperidine-1-carboximidamide hydroiodide
Videos
SDS
Documents

Documents

Product Certifications