missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYHIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PHYHIP |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816794
|
Proteintech
16984-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806794
|
Proteintech
16984-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes both PHYHIP (35-38 kDa) and PHYHIPL (43-45 kDa).
Its interaction with PHYH suggests a role in the development of the central system.Specifications
PHYHIP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q568Z9, Q8K0S0, Q92561 | |
105653, 290356, 9796 | |
PHYHIP Fusion Protein Ag10575 | |
Primary | |
-20°C | |
PHYHIP |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DYRK1AP3, KIAA0273, PAHX AP, PAHX AP1, PAHXAP1, PHYHIP | |
Phyhip | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |