missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic anhydride in pyridine, BAKER ANALYZED™, J.T. Baker™
49.17 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.
401.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "21637" to get your promotional price.
401.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Click to view available options
2.5 L
Specifications
Color | Max. 100 (APHA) |
Flash Point | 17°C |
Packaging | Glass container |
Molecular Formula | C8H4O3 |
MDL Number | MFCD00005918 |
Quantity | 2.5 L |
Molecular Weight (g/mol) | 148.12 |
Concentration or Composition (by Analyte or Components) | 160 g/L |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'