missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus lump, red, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

Specifications

CAS 7723-14-0
Physical Form Solid
Percent Purity ≥99.999%
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7723-14-0
Solid
≥99.999%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phosphorus lump, red

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H228-Flammable solid.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications