missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Smtnl1 (Ser301) Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19560-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognize Ser(301) phospho of Smtn1.
Plays a role in the regulation of contractile properties of both striated and smooth muscles. When unphosphorylated, may inhibit myosin dephosphorylation. Phosphorylation at Ser-299 reduces this inhibitory activity.Specifications
Phospho-Smtnl1 (Ser301) | |
Polyclonal | |
Unconjugated | |
Smtnl1 | |
1110030K22Rik, CHASM, smoothelin like 1, Smtnl1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68678 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99LM3 | |
Smtnl1 | |
Peptide | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |