missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Smtnl1 (Ser301) Rabbit anti-Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19560-1-AP-100UL

 View more versions of this product

Product Code. 16889094

  • £256.47 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The antibody recognize Ser(301) phospho of Smtn1.

Plays a role in the regulation of contractile properties of both striated and smooth muscles. When unphosphorylated, may inhibit myosin dephosphorylation. Phosphorylation at Ser-299 reduces this inhibitory activity.
Specifications

Specifications

Phospho-Smtnl1 (Ser301)
Polyclonal
Unconjugated
Smtnl1
1110030K22Rik, CHASM, smoothelin like 1, Smtnl1
Rabbit
Antigen Affinity Chromatography
RUO
68678
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99LM3
Smtnl1
Peptide
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers