missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-FANCD2 (Ser330) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19543-1-AP-100UL
This item is not returnable.
View return policy
Description
The antibody is specific to phospho-Ser(330) of FANCD2.
Fanconi anemia (FANC) is a human autosomal-recessive cancer susceptibility disorder characterized by congenital defects, progressive bone marrow failure, and cellular hypersensitivity to mitomycin C (MMC). The FANC subunit D2 protein is vital for cellular resistance to DNA cross-linking and the arrest of DNA synthesis after ionizing radiation. DNA damage activates the monoubiquitination of FANCD2, targeting nuclear foci containing the BRCA1 protein.Specifications
Phospho-FANCD2 (Ser330) | |
Polyclonal | |
Unconjugated | |
FANCD2 | |
FA D2, FA4, FACD, FAD, FAD2, FANCD, FANCD2, Protein FACD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2177 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BXW9 | |
FANCD2 | |
Peptide | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |