missing translation for 'onlineSavingsMsg'
Learn More

PBS with Tween™ 20, Phosphate Buffered Saline, Ready-to-Dissolve pouches, pH 7.4, Fisher BioReagents

Brand:  Fisher Bioreagents BP2938-10

Code : 32

Additional Details : Weight : 0.01000kg

Product Code. 10869020

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Contents of pouch dissolved in 1L ultrapure water yield a 0.01M phosphate buffered saline solution
  • Quick and easy to mix, dissolution time is less than 5 minutes
  • 1L solution contains 0.138M NaCl, 0.0027M KCl, and 0.05% Tween 20
  • Sold as 10 foil pouches each containing 10g of preweighed content
  • PBS-Tween solutions are used as wash buffers and diluent for immunodetection techniques
  • The addition of Tween 20, a detergent, helps to decrease non-specific binding and improve detection
  • Intended for laboratory research use only
Specifications

Specifications

Phosphate Buffered Saline
Powder/Solid
PBS-Tween
10g x 10 foil pouches
7.4
Videos
SDS Documents
Certificates