missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethylammonium tribromide, 97%, Thermo Scientific Chemicals

Catalog Number p-7022507
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
4207-56-1
C9H14I3N
516.931
MFCD00011789
RHXJPCQWYPXLEH-UHFFFAOYSA-N
mono n,n,n-trimethylbenzenaminium tribromide
21525000
trimethyl(phenyl)azanium;triiodide
C[N+](C)(C)C1=CC=CC=C1.I[I-]I
This item is not returnable. View return policy
4207-56-1
C9H14I3N
516.931
MFCD00011789
RHXJPCQWYPXLEH-UHFFFAOYSA-N
mono n,n,n-trimethylbenzenaminium tribromide
21525000
trimethyl(phenyl)azanium;triiodide
C[N+](C)(C)C1=CC=CC=C1.I[I-]I

Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium tribromide is used as brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents, reducing agents and bases.
TRUSTED_SUSTAINABILITY
Melting Point 112°C to 117°C
Odor Irritating
Quantity 50 g
UN Number UN3261
Beilstein 3919459
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 375.93
Percent Purity 97%
Chemical Name or Material Phenyltrimethylammonium tribromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'