missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM1 Mouse anti-Human, Clone: 2G5A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66105-1-IG-20UL
This item is not returnable.
View return policy
Description
Phosphoglucomutases.Specifications
PGM1 | |
Monoclonal | |
0.82 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P36871 | |
PGM1 | |
PGM1 Fusion Protein Ag7294 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
2G5A8 | |
Unconjugated | |
PGM1 | |
Glucose phosphomutase 1, PGM 1, PGM1, phosphoglucomutase 1 | |
Mouse | |
Protein A | |
RUO | |
5236 | |
-20°C | |
Liquid |