missing translation for 'onlineSavingsMsg'
Learn More

PGM1 Mouse anti-Human, Clone: 2G5A8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66105-1-IG-150UL

 View more versions of this product

Product Code. 16877675

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Phosphoglucomutases.
Specifications

Specifications

PGM1
Monoclonal
0.82 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P36871
PGM1
PGM1 Fusion Protein Ag7294
150 μL
Primary
Human
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2G5A8
Unconjugated
PGM1
Glucose phosphomutase 1, PGM 1, PGM1, phosphoglucomutase 1
Mouse
Protein A
RUO
5236
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers