missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Honeywell™ Riedel-de Haën™

Catalog Number 15623780
386.67 GBP valid until 2024-03-31
Use promo code "21663" to get your promotional price.


Click to view available options
Quantity:
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
101316-46-5
MFCD00081849
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
101316-46-5
MFCD00081849

Puriss. p.a., ACS Reagent, Reag. ISO, low boiling point hydrogen treated naphtha, bp≥ 90% 40-60 °C (≥ 90%)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Appearance Liquid
UN Number UN1268
Grade ACS/puriss. p.a.
Density 0.640 to 0.655g/mL at 20°C
Boiling Point 40°C to 60°C
Quantity 4 x 2.5 L
Flash Point −30°C (−22°F)
Packaging Glass Bottle
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid (as CH3COOH): ≤0.002%
Chemical Name or Material Petroleum Ether, Reag. ISO
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C6, Isoalkanes, <5% n-Hexane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'