missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, low boiling point, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Specifications
Specifications
Density | 0.63g/mL at 20°C |
Boiling Point | 30°C to 40°C |
Quantity | 500 mL |
Flash Point | −49°C (−56.2°F) |
UN Number | UN1268 |
Chemical Name or Material | Petroleum ether |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'