missing translation for 'onlineSavingsMsg'
Learn More

3M™ Petrifilm Aerobic Count Plates

£118.20 - £134.00

Specifications

For Use With (Application) Determines the population of aerobic bacteria
Host Species Lactic-acid bacteria
Specimen Type Lactic-acid Bacteria
Type Petrifilm Aerobic Count Plates
Species Lactic-acid bacteria
View More Specs

Products 2
Product Code Brand Packaging Price Quantity & Availability  
Product Code Brand Packaging Price Quantity & Availability  
10044574
View Documents
3M™
7100039310
100 Pk.
£118.20
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 24-09-2021
Add to basket
 
12842191
View Documents
3M™
7100039310
100 Pk. £134.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 18-08-2021
Add to basket
 
Description

Description

Specifications

Specifications

Determines the population of aerobic bacteria
Lactic-acid Bacteria
Lactic-acid bacteria
Lactic-acid bacteria
Petrifilm Aerobic Count Plates
SDS
Product Certifications