missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE7A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26277-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE7 subfamily. This PDE hydrolyzes the second messenger, cAMP, which is a regulator and mediator of a number of cellular responses to extracellular signals. Thus, by regulating the cellular concentration of cAMP, this protein plays a key role in many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
PDE7A | |
Polyclonal | |
Unconjugated | |
PDE7A | |
HCP1, PDE7, PDE7A, phosphodiesterase 7A, TM22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5150 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13946 | |
PDE7A | |
PDE7A Fusion Protein Ag24334 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |