Learn More
PARD3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11085-1-AP-150UL
This item is not returnable.
View return policy
Description
The PARD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PARD3. This antibody recognizes human, mouse, rat antigen. The PARD3 antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
PARD3 | |
Polyclonal | |
Unconjugated | |
150 μL | |
56288 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |