missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyltrichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11440337
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-35-9
Molecular Formula C7H7Cl3Si
Molecular Weight (g/mol) 225.568
MDL Number MFCD00045131
InChI Key WOMUGKOOLXQCTQ-UHFFFAOYSA-N
Synonym p-tolyltrichlorosilane, tolyltrichlorosilane, trichloro-p-tolylsilane, silane, trichloro 4-methylphenyl, trichloro 4-methylphenyl silane, trichloro p-tolyl silane, silane, trichloro-p-tolyl, benzene, 1-methyl-4-trichlorosilyl, 4-tolyltrichlorosilane, p-tolylsilyl trichloride
PubChem CID 69697
IUPAC Name trichloro-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 218°C to 220°C
Quantity 25 g
UN Number UN2987
Beilstein 1866557
Flash Point 98°C (208°F)
Formula Weight 225.58
Percent Purity 97%
Odor Irritating
Chemical Name or Material p-Tolyltrichlorosilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'