missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement
Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.
£140.00 - £539.00
Specifications
Species | Legionella |
---|---|
Description | Legionella BCYE Supplement |
Form | Freeze-dried |
Product Type | Microbiology Media |
Quantity | 10/Pk. |
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
17180661
|
Thermo Scientific™
SR0251A |
Each vial for 100mL medium |
£140.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
17190661
|
Thermo Scientific™
SR0251C |
Each vial for 500mL medium |
£539.00
Pack of 10 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
Description
Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.
Add to Legionella Agar Base, Part No. CM1203BSpecifications
Legionella | |
Freeze-dried | |
10/Pk. |
Legionella BCYE Supplement | |
Microbiology Media |