Learn More
Thermo Scientific™ Antibiotic Medium No. 3 Assay Broth (Dehydrated)
Carry out serial dilution assay of penicillin and other antibiotics with this medium.
Brand: Thermo Scientific™ CM0287B
141.02 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Carry out serial dilution assay of penicillin and other antibiotics with Thermo Scientific™ Oxoid™ Antibiotic Medium No. 3 Assay Broth (Dehydrated). Antibiotic Medium No. 3 Assay Broth is used as the broth medium in turbidimetric or serial dilution assay of a wide variety of antibiotics with Staphylococcus aureus.
Antibiotic Medium No. 3 Assay Broth is used as the broth medium in turbidimetric or serial dilution assay of a wide variety of antibiotics with Staphylococcus aureus.
Taking advantage of modern technology, this medium is based on the original `Penassay Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Antibiotic Medium No. 3 | |
Dehydrated Culture Media | |
For 28.6L medium |
Powder | |
500 g |