missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Alternative Thioglycollate Broth
Oxoid Thioglycollate Broth, Alternative is a non-viscous thioglycollate medium is used for the culture of turbid biological products.
Brand: Thermo Scientific™ CM0391B
169.41 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Oxoid Thioglycollate Broth, Alternative is a non-viscous thioglycollate medium is used for the culture of turbid biological products.
Specifications
Industrial Reference: USP Alternative. | |
Powder | |
500 g |
Thioglycollate Broth (USP alternative) | |
Dehydrated Culture Media | |
For 17.2L medium |
For Laboratory Use Only