missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073351
123.69 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


74.98 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
112-80-1
C18H34O2
282.47
MFCD00064242
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
445639
CHEBI:16196
(Z)-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O
112-80-1
C18H34O2
282.47
MFCD00064242
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
445639
CHEBI:16196
(Z)-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O

C18H34O2, CAS Number-112-80-1, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, glycon ro, glycon wo, oleate, oleic acid, pamolyn 100, wecoline oo, z-octadec-9-enoic acid, 2.5L, 360 deg.C, CHEBI:16196, Colorless, 282.46g/mol, ZQPPMHVWECSIRJ-KTKRTIGZSA-N, (Z)-octadec-9-enoic acid, 13 deg.C

TRUSTED_SUSTAINABILITY
Melting Point 13°C
Color Colorless
Boiling Point 360°C
Packaging Amber glass bottle
Quantity 1 L
Vapor Density 9.7
Viscosity 39.1 mPaS at 20°C
Formula Weight 282.46g/mol
Vapor Pressure 1mmHg 176
Physical Form Liquid
compliance-icons
Product Identifier
  • Oleic acid
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'