missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODF3 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25097-1-AP-20UL
This item is not returnable.
View return policy
Description
Outer dense fibers are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail. May help to maintain the passive elastic structures and elastic recoil of the sperm tail.Specifications
ODF3 | |
Polyclonal | |
Unconjugated | |
ODF3 | |
hSHIPPO, ODF3, Outer dense fiber protein 3, SHIPPO1, Sperm tail protein SHIPPO 1, TISP50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113746, 365387 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96PU9 | |
Odf3 | |
ODF3 Fusion Protein Ag18734 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |