missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octan-1-ol, Pure, Fisher Chemical™
68.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
68.18 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
68.18 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Specifications
Melting Point | -16°C |
Color | Colorless |
Boiling Point | 195°C |
Packaging | Amber glass bottle |
Quantity | 500 mL |
Vapor Density | 4.5 |
Formula Weight | 130.23g/mol |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'