missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP133 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12405-1-AP-20UL
This item is not returnable.
View return policy
Description
Plays a role in T-cell signaling pathway. Isoform 2 may play a role in T-helper 2 (Th2) signaling pathway and seems to represent the first proximal signaling protein that links T-cell receptor-mediated signal to the activation of c-Maf Th2 specific factor.Specifications
NUP133 | |
Polyclonal | |
Unconjugated | |
NUP133 | |
133 kDa nucleoporin, hNUP133, nucleoporin 133kDa, Nucleoporin Nup133, NUP133 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234865, 292085, 55746 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R0G9, Q8WUM0 | |
NUP133 | |
NUP133 Fusion Protein Ag3100 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |