missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoBond™ Buffer Set IV

Buffer set for the isolation of genomic DNA from tissue using NucleoBond AXG columns; sufficient for 10 AXG 500 columns (Maxi preps), 20 AXG 100 columns (Midi preps), or 100 AXG 20 columns (Mini preps).

Brand:  Macherey-Nagel Bioanalysis™ 740604


Product Code. 10722577

  • £134.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Include all the required buffers and enzymes (proteinase K and RNase A) for the extraction of genomic DNA from tissue and bacteria. Using these buffer sets in combination with NucleoBond™ AXG columns allows isolation of up to 500 μg genomic DNA from different matrices.

The quality of the genomic DNA is extraordinarily high and is suitable for all kinds of subsequent reactions. NucleoBond™ AXG columns run by gravity flow resulting in a high molecular weight DNA without shearing. Samples are first lysed with proteinase K. Before loading the lysate to the NucleoBond™ AXG column, it is diluted with buffer N2 in order to guarantee a good flow rate as well as a quantitative binding of the DNA. After efficient washing the genomic DNA is eluted with high-salt buffer and precipitated with isopropanol. NucleoBond™ AXG columns are available in three sizes (AXG 20, AXG 100 and AXG 500).NucleoBond™ buffer sets:NucleoBond™ buffer set III for the isolation of genomic DNA from bacteria: for 10 AXG 500 columns (maxi preps), 20 AXG 100 columns (midi preps) or 100 AXG 20 columns (mini preps).NucleoBond™ buffer set IV for the isolation of genomic DNA from tissue: for 10 AXG 500 columns (maxi preps), 20 AXG 100 columns (midi preps) or 100 AXG 20 columns (mini preps).For further information on this product please contact Customer Services.

  • NucleoBond™ AXG columns with NucleoBond™ buffer set III for genomic DNA from bacteria and yeast (for yeast additional buffers required)
  • NucleoBond™ AXG columns with NucleoBond™ buffer set IV for genomic DNA from tissue
  • Anion exchange chromatography
  • Sample size: up to 80 mL cell culture or up to 400 mg tissue
  • Yield: up to 500 μg genomic DNA
  • Fast and versatile separation system
  • Columns run by gravity flow
  • High batch-to-batch reproducibility
  • No use of organic solvents
  • Ultra-pure nucleic acids suitable for all downstream applications
Specifications

Specifications

18°C to 25°C
Set of 125 mL buffers G2, N2, N3, N5, Proteinase K and Rnase
Buffer
Isolation of DNA
1 Set
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers