Learn More
Invitrogen™ Luminex™ IL-5 Human Singleplex Bead Kit
Analyte specific components for the measurement of human IL-5 (Interleukin-5) in serum, plasma or tissue culture supernatant.
Brand:  Invitrogen™ LHC0051
Code : 50
Additional Details : Weight : 0.06000kg
Product Code. 10120473
  • £253.00 / 100 tests
Estimated Shipment: 07-07-2021
Add to basket
For Research Use Only. All usage must comply with product instructions.
Includes: Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.
Description
Description
Assay may be run alone or in combination with other Human Singleplex Bead Kits from Invitrogen. EveryPrep™ Universal Vacuum Manifold available separately (Cat. No. K2111-01). Reagents intended for use in Luminex 100™ or 200™ Systems only.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Specifications
NM_000879.2, NP_000870.1
Fluorescent Detection
Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard
interleukin 5 (colony-stimulating factor, eosinophil)
Serum
Luminex Instruments
Polystyrene
3567
IL-5
100 Assays
<3pg/mL