missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Novagen™ Benzonase™ Nuclease

Used for removal of nucleic acids from recombinant proteins

Brand:  Merck Millipore 70746-3

Code : NEW

Additional Details : Weight : 0.01080kg

Product Code. 10349963

  • £185.00 / Each

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Genetically engineered endonuclease from Serratia marcescens
  • Degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity
  • Effective over wide range of conditions and possesses high specific activity
  • Rapidly hydrolyzes nucleic acids to makes enzyme for viscosity reduction to reduce processing time and increase yields of protein
  • Compatible with BugBuster™ and PopCulture™ Protein Extraction Reagents
  • Can be added along with these reagents to eliminate viscosity and remove nucleic acids from E. coli extracts
  • Functional between pH 6 and 10 and from 0 to 42°C and requires 1to 2mM Mg2+ for activation
  • Active in presence of ionic and non-ionic detergents, reducing agents, PMSF (1mM), EDTA (1mM) and urea
  • Activity is inhibited by >150mM monovalent cations, >100mM phosphate, >100mM ammonium sulfate, or >100mM guanidine HCl
  • Packaging: Plastic bottle

Compliance

Compliant with FDA

Specifications

Specifications

25 to 29U/μL
Supplied in 50% glycerol
>90%
-20°C
Nuclease
Nucleic acids removal from recombinant proteins
6 to 10
10ku
RUO
Videos
SDS
Documents
Certificates