missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonane, 99%, Reagent Grade, Honeywell™
Reagent Grade, 99%
Chemical Identifiers
CAS | 111-84-2 |
---|---|
Molecular Formula | C9H20 |
Molecular Weight (g/mol) | 128.259 |
MDL Number | MFCD00009574 |
InChI Key | BKIMMITUMNQMOS-UHFFFAOYSA-N |
Synonym | n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard |
PubChem CID | 8141 |
ChEBI | CHEBI:32892 |
IUPAC Name | nonane |
SMILES | CCCCCCCCC |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
111-84-2 | |
128.259 | |
BKIMMITUMNQMOS-UHFFFAOYSA-N | |
8141 | |
nonane |
C9H20 | |
MFCD00009574 | |
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard | |
CHEBI:32892 | |
CCCCCCCCC |
Specifications
111-84-2 | |
Colorless | |
151°C (1013 hPa) | |
Characteristic | |
CH3(CH2)7CH3 | |
UN1920 | |
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard | |
CCCCCCCCC | |
128.259 | |
CHEBI:32892 | |
0.99 | |
Nonane |
-53°C | |
0.718g/mL at 25°C | |
31°C (87.8°F) | |
C9H20 | |
MFCD00009574 | |
1696917 | |
BKIMMITUMNQMOS-UHFFFAOYSA-N | |
nonane | |
8141 | |
128.26g/mol | |
Liquid |
Safety and Handling
P210-P273-P301 + P310-P304 + P340 + P312-P331-P391
missing translation for 'dotInformation' : 3/PGIII
missing translation for 'rtecsNumber' : 203-913-4