missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rogo Sampaic™ Inoculation Loops without Eyelets
For retrieving inoculums from a culture of microorganisms. Rogo Sampaic™ Inoculation Loops without Eyelets provide a superior solution for transferring microscopic microorganisms, inoculums or for streaking.
Brand: Rogo Sampaic™ BRG502
Description
- 290mm long
- Manufactured from aluminum and nickel
- Bare handle without eyelet
Specifications
290 mm | |
Nickel | |
Nickel Handle | |
1 Ea. |